missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, anhydrous, 99% (metals basis), Thermo Scientific Chemicals

121.61 € - 521.60 €

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11356998
View Documents
Thermo Scientific Alfa Aesar
014687.22
100 g
121.61 €
100g
Please to purchase this item. Need a web account? Register with us today!
11346998
View Documents
Thermo Scientific Alfa Aesar
014687.36
500 g
521.60 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel(II) chloride is a mild Lewis acid used for the regioselective isomerization of dienols and selective reduction of alkenes to alkanes in presence of Lithium aluminum hydride. It is used as a precursor to nickel boride, finely divided nickel. It acts as a catalyst to prepare dialkyl arylphosphonates from phosphites and aryl iodide. It finds application in the electroplating of nickel onto other metal items. It is also used in the synthesis of semiconducting metal-containing polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7718-54-9
129.593
QMMRZOWCJAIUJA-UHFFFAOYSA-L
CHEBI:34887
Cl[Ni]Cl
Cl2Ni
MFCD00011142
24385
dichloronickel

Specifications

1001°C
Yellow to Orange
99% (metals basis)
MFCD00011142
QMMRZOWCJAIUJA-UHFFFAOYSA-L
dichloronickel
24385
129.6
Yellow to orange powder
3.55 g/cm3
7718-54-9
Powder
Cl2Ni
Soluble in water,ethylene glycol,ethanol and ammonium hydroxide. Insoluble in ammonia and nitric acid.
Cl[Ni]Cl
129.593
CHEBI:34887
Hygroscopic
1.33 hPa (1mm Hg) at 20°C
Nickel(II) chloride, anhydrous
Videos
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c

H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500

missing translation for 'dotInformation' : Hazard Class: 6.1, Packing Group: III

missing translation for 'einecsNumber' : 231-743-0

missing translation for 'rtecsNumber' : QR6475000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only