missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11356937
Click to view available options
Quantity:
5 g
25 g
100 g

It is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69098-15-3
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.59
MDL Number MFCD00149808
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
Synonym nickel ii chloride hydrate, nickel chloride hydrate, cl2ni.h2o, nicl2.h2o, nickel 11 chloride hydrate, nickel ii chloride hydrate, puratronic, nickel ii chloride hydrate trace metals basis
PubChem CID 16212333
SMILES [Cl-].[Cl-].[Ni++]

Specifications

Color Green
Physical Form Crystalline Aggregates
Quantity 5 g
Linear Formula NiCl2·xH2O
UN Number UN3288
Merck Index 14,6505
Solubility Information Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide.
IUPAC Name dichloronickel;hydrate
Formula Weight 129.60 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Nickel(II) chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'