missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

152.76 € - 1513.53 €

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11348557
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035713.06
5 g
152.76 €
5g
Please to purchase this item. Need a web account? Register with us today!
11368557
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035713.14
25 g
559.79 €
25g
Please to purchase this item. Need a web account? Register with us today!
11358557
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035713.22
100 g
1513.53 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7718-54-9
129.593
QMMRZOWCJAIUJA-UHFFFAOYSA-L
CHEBI:34887
Cl[Ni]Cl
Cl2Ni
MFCD00011142
24385
dichloronickel

Specifications

1001°C
Yellow to Orange
99.9% (metals basis)
MFCD00011142
QMMRZOWCJAIUJA-UHFFFAOYSA-L
dichloronickel
24385
129.6
Hygroscopic
1.33 hPa (1mm Hg) at 20°C
Nickel(II) chloride
7718-54-9
Powder
Cl2Ni
Soluble in water.
Cl[Ni]Cl
129.593
CHEBI:34887
Odorless
Yellow to orange powder
3.55
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c

H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500

missing translation for 'dotInformation' : DOT Class: 6.1; Packing Group: III

missing translation for 'einecsNumber' : 231-743-0

missing translation for 'rtecsNumber' : QR6475000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only