missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) hydroxide, Ni 61%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Description
Due to its redox behavior, it finds use in rechargeable battery electrodes. It is useful for the storage of electrochemical energy, with potential applications in car batteries. It is a potential material for several biological applications, for example in chromosomal DNA quantification assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 230°C (decomposition) |
Color | Green |
Physical Form | Powder |
Quantity | 250 g |
Linear Formula | Ni(OH)2 |
UN Number | UN3077 |
Solubility Information | Insoluble in water. Soluble in dilute acid. |
Formula Weight | 92.7 |
Concentration or Composition (by Analyte or Components) | Ni 61% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'