missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) iodide, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals

56.58 € - 189.95 €

Chemical Identifiers

CAS 13462-90-3
Molecular Formula I2Ni
Molecular Weight (g/mol) 312.502
MDL Number MFCD00016264
InChI Key BFSQJYRFLQUZKX-UHFFFAOYSA-L
Synonym nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
PubChem CID 26038
IUPAC Name diiodonickel
SMILES [Ni](I)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358117
View Documents
Thermo Scientific Alfa Aesar
022893.06
5 g
56.58 €
5g
Please to purchase this item. Need a web account? Register with us today!
11348117
View Documents
Thermo Scientific Alfa Aesar
022893.14
25 g
189.95 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel(II) iodide serves as a catalyst in carbonylation reactions. It is also used as a reagent in organic synthesis, especially in conjunction with samarium(II) iodide. It plays an important role in homogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13462-90-3
312.502
BFSQJYRFLQUZKX-UHFFFAOYSA-L
26038
[Ni](I)I
I2Ni
MFCD00016264
nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
diiodonickel

Specifications

797°C
I2Ni
UN3077
Soluble in water and alcohols.
[Ni](I)I
312.502
312.51
Odorless
5.834 g/mL
Nickel(II) iodide, Anhydrous
13462-90-3
MFCD00016264
nickel ii iodide, nickel iodide, nickel iodide nii2, nickel diiodide, nickel 2+ iodide, nii2, acmc-1bxz2, nickel ii iodide, powder
BFSQJYRFLQUZKX-UHFFFAOYSA-L
diiodonickel
26038
99.5%
Hygroscopic; Light sensitive
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H341-H350i-H360D-H372

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 236-666-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Nickel(II) iodide, anhydrous

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : Nickel iodide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.