missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) Oxide, Pure, Black, Fisher Chemical Product Code.: 10141960

Nickel(II) Oxide, Pure, Black, Fisher Chemical

Product Code. 10141960
500 g, Plastic powder jar
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10141960

Brand: Fisher Chemical N/1850/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 1960°C
CAS 1313-99-1
Color Black
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula NiO
MDL Number MFCD00011145
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILES O=[Ni]
IUPAC Name oxonickel
Molecular Weight (g/mol) 74.69
PubChem CID 14805
Formula Weight 74.7g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) oxide
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Carcinogenicity Category 1A
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H350i-May cause cancer by inhalation.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) Oxide, Pure, Black, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.