missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) perchlorate hexahydrate, Reagent Grade

Product Code. 11371728
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11371728

Brand: Thermo Scientific Alfa Aesar 011631.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13520-61-1 | Cl2H12NiO14 | 365.675 g/mol

Nickel(II) perchlorate hexahydrate is used as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13520-61-1
Molecular Formula Cl2H12NiO14
Molecular Weight (g/mol) 365.675
MDL Number MFCD00149804
InChI Key WHGYCGOFTBFDLW-UHFFFAOYSA-L
Synonym nickel ii perchlorate hexahydrate, acmc-1boln, 2clo4.ni.6h2o, nickel ii perchlorate hydrate, nickel 2+ hexahydrate diperchlorate, nickel perchlorate, hexahydrate, reagent, nickel ii perchlorate hexahydrate, reagent grade, nickel ii perchlorate hexahydrate trace metals basis, nickel ii perchlorate hexahydrate, purum p.a., crystallized kt
PubChem CID 15769463
IUPAC Name nickel(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2]

Specifications

Melting Point 140°C
Color Green
Physical Form Damp Crystalline Solid
Quantity 500 g
Solubility Information Very soluble in water. Soluble in many organic solvents
Formula Weight 365.70 (257.59 Anhydrous)
Grade Reagent
Odor Odorless
Refractive Index 1.55
Sensitivity Hygroscopic
Appearance Green crystals
Chemical Name or Material Nickel(II) perchlorate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.