missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) phthalocyanine, Thermo Scientific Chemicals

Product Code. 11378248
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14055-02-8
Molecular Formula C32H16N8Ni
Molecular Weight (g/mol) 571.23
MDL Number MFCD00041954
InChI Key LVIYYTJTOKJJOC-UHFFFAOYSA-N
Synonym nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 %
PubChem CID 2735100
IUPAC Name nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

Specifications

Melting Point >300°C
Color Blue to Purple
Quantity 5 g
Formula Weight 571.24
Physical Form Powder
Chemical Name or Material Nickel(II) phthalocyanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.