missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfamate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Nickel plating

49.75 € - 165.83 €

Chemical Identifiers

CAS 13770-89-3
Molecular Formula H4N2NiO6S2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00137261
InChI Key KERTUBUCQCSNJU-UHFFFAOYSA-L
Synonym nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
PubChem CID 83720
IUPAC Name nickel(2+);disulfamate
SMILES [Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11308488
View Documents
Thermo Scientific Alfa Aesar
040501.K2
1 L
49.75 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11450281
View Documents
Thermo Scientific Alfa Aesar
040501.8M
2 x 2.5 L
165.83 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel plating Nickel(II) sulfamate is used for electroplating and as a surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13770-89-3
250.85
KERTUBUCQCSNJU-UHFFFAOYSA-L
83720
[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
H4N2NiO6S2
MFCD00137261
nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
nickel(2+);disulfamate

Specifications

13770-89-3
Liquid
Ni(SO3NH2)2
UN3082
Miscible with water,immiscible with ether,acetone.
[Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O
250.85
250.86 (Anhydrous)
50% w/w aq. soln.
Nickel(II) sulfamate
Green
H4N2NiO6S2
MFCD00137261
nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
KERTUBUCQCSNJU-UHFFFAOYSA-L
nickel(2+);disulfamate
83720
Reagent
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H341-H350i-H360D-H372

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 237-396-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only