missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfamate hydrate, Thermo Scientific Chemicals

Product Code. 11349369
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13770-89-3
Molecular Formula H4N2NiO6S2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00137261
InChI Key KERTUBUCQCSNJU-UHFFFAOYSA-L
Synonym nickel sulfamate, nickel bis sulphamidate, nickel ii sulfamate, aeronikl 250, aeronikl 400, aeronikl 575, nickel 2+ disulfamate, sulfamic acid, nickel 2+ salt 2:1, nickel sulfamate 6ci,7ci
PubChem CID 83720
SMILES [Ni++].NS([O-])(=O)=O.NS([O-])(=O)=O

Specifications

Color Blue
Physical Form Crystalline
Quantity 250 g
UN Number UN3077
Solubility Information Soluble in water.
IUPAC Name nickel(2+);disulfamate
Formula Weight 250.86 (Anhydrous)
Odor Odorless
Chemical Name or Material Nickel(II) sulfamate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'