missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) sulfate, 0.5M aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
100 mL
Description
Nickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.
Solubility
Fully miscible in water.
Notes
Protect from heat.
Nickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.
Solubility
Fully miscible in water.
Notes
Protect from heat.
Specifications
Specifications
Color | Green |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | NiSO4 |
UN Number | UN3082 |
Solubility Information | Fully miscible in water. |
Formula Weight | 262.86 |
Concentration or Composition (by Analyte or Components) | 0.5M aq. soln. |
Odor | Odorless |
Chemical Name or Material | Nickel(II) sulfate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'