Learn More
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
28.40 € - 458.00 €
Chemical Identifiers
CAS | 10101-97-0, 7786-81-4 |
---|---|
Molecular Formula | NiO4S·6H2O |
Molecular Weight (g/mol) | 262.85 |
MDL Number | MFCD00149813 |
InChI Key | RRIWRJBSCGCBID-UHFFFAOYSA-L |
Synonym | nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 |
PubChem CID | 5284429 |
ChEBI | CHEBI:53437 |
IUPAC Name | nickel(2+);sulfate;hexahydrate |
SMILES | O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10034013
|
Thermo Scientific Acros
415611000 |
100 g | Plastic bottle |
28.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10361681
|
Thermo Scientific Acros
415615000 |
500 g | Plastic bottle |
79.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10155123
|
Thermo Scientific Acros
415610010 |
1 kg | Plastic bottle |
134.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13408568
|
Thermo Scientific Acros
415610051 |
5 kg | Plastic Bottle |
458.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10101-97-0, 7786-81-4 | |
262.85 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
5284429 | |
nickel(2+);sulfate;hexahydrate |
NiO4S·6H2O | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
CHEBI:53437 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Specifications
53.0°C | |
Blue-Green | |
100.0 | |
98.0 to 102% (Complexometry) | |
NiSO4·6H2O | |
15,6602 | |
Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
262.85 | |
CHEBI:53437 | |
≥98% | |
Nickel(II) sulfate hexahydrate |
10101-97-0, 7786-81-4 | |
98.0 | |
Crystals | |
NiO4S·6H2O | |
MFCD00149813 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
nickel(2+);sulfate;hexahydrate | |
5284429 | |
262.85 | |
ACS Reagent |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Suspected of causing genetic defects.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
M
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : QR9600000
- Nickel(II) sulfate hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only