missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Sulfate Hexahydrate, ≥99.99% Trace Metals Basis, Honeywell™

Product Code. 15685620
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

≥99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Melting Point 53°C
Color Green
pH 4 to 6 (at 20°C)
Physical Form Solid
Quantity 50 g
Linear Formula NiSO4 · 6H2O
UN Number UN3077
Solubility Information In water 378g/L at 20°C
Formula Weight 262.85g/mol
Percent Purity ≥99.99% (trace metals basis)
Odor Odorless
Packaging Poly Bottle
Density 1.95g/cm3; Vapor Density: 2.07 (vs air)
Chemical Name or Material Nickel(II) sulfate hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'