missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) sulfate hexahydrate, Puriss. p.a., ACS Reagent, 99-102.0%, Honeywell Fluka™

Product Code. 15693460
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, 99-102.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-97-0
Molecular Formula H12NiO10S
Molecular Weight (g/mol) 262.839
MDL Number MFCD00149813
InChI Key RRIWRJBSCGCBID-UHFFFAOYSA-L
Synonym nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928
PubChem CID 5284429
ChEBI CHEBI:53437
IUPAC Name nickel(2+);sulfate;hexahydrate
SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]

Specifications

Quantity 100 g
Linear Formula NiSO4 · 6H2O
UN Number 3077
Formula Weight 262.85g/mol
Percent Purity 99 to 102%
Grade ACS/puriss. p.a.
Density Vapor Density: 2.07 (vs air)
Chemical Name or Material Nickel(Ii) Sulfate Hexahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'