missing translation for 'onlineSavingsMsg'
Learn More

Nickel phthalocyanine, 95%, Thermo Scientific Chemicals

Product Code. 10015690
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14055-02-8
Molecular Formula C32H16N8Ni
Molecular Weight (g/mol) 571.23
MDL Number MFCD00041954
InChI Key LVIYYTJTOKJJOC-UHFFFAOYSA-N
Synonym nickel phthalocyanine, nickel ii phthalocyanine, phthalocyanine, ni, nickel ii phthalocyanine, dye content 85 %
PubChem CID 2735100
IUPAC Name nickel(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Ni++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Blue
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 571.21
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Nickel phthalocyanine, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Nickel phthalocyanine
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'