missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Nickel-Plated Brass Open Armor Thermometer Cases

Feature screw cap with ring and small openings around bulb for circulation

Brand:  H-B Instrument™ B61400-9300

 View more versions of this product

Product Code. 17571305

  • 30.65 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Can hold thermometers of length 5.9 to 18 in. (150mm to 460mm)

Specifications

Specifications

Protects thermometer during use and storage
Nickel-plated, Brass
12 in.
300 mm
Armor Case
12 in. Thermometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers