missing translation for 'onlineSavingsMsg'
Learn More

Usbeck Laborgeraete™ Nickel-plated Clamps with Vinyl-coated Jaws

Secure delicate glassware in the laboratory with Usbeck Laborgeraete™ Nickel-plated Clamps. The double-sided opening mechanism allows jaws to spread evenly for stabilization and balance. .

Brand:  Usbeck Laborgeraete™ 2544

 View more versions of this product

Product Code. 10208821

  • 15.40 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Zinc diecasting
  • Powdercoated
  • Nickel plated vinyl coated jaws
Specifications

Specifications

1 Ea.
Nickel-plated
235 mm
Clamp
12 to 25 mm
Vinyl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers