Learn More
Nickel plating solution, electroless, Thermo Scientific Chemicals
For Plating On Silicon, Germanium, Gallium Arsenide
Brand:  Thermo Scientific Alfa Aesar 044069.AK
 View more versions of this product
Product Code. 11375148
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For plating on silicon, germanium, gallium arsenide, cadmium sulfide, gold, nickel, iron and Kovar
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nickel plating solution
Green
Ammonia-like
Liquid
250 mL
MFCD00011137