missing translation for 'onlineSavingsMsg'
Learn More
Nickel powder, -100 mesh, 99+% (metals basis excluding Co), Co 0.1% max, Thermo Scientific Chemicals Product Code.: 11340699

Nickel powder, -100 mesh, 99+% (metals basis excluding Co), Co 0.1% max, Thermo Scientific Chemicals

Product Code. 11340699
250 g
Click to view available options
Quantity:
25 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11340699

Brand: Thermo Scientific Alfa Aesar 000224.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 250 g
Percent Purity ≥99%
Mesh Size −100 Mesh
Odor Odorless
Form Powder
Assay (metals basis excluding Co), Co 0.1% max.
Chemical Name or Material Nickel powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX23A026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000224.30,000224.A1,000224.14
Lot NumberF16U012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000224.30,000224.A1,000224.14
Lot NumberS22H026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000224.30,000224.A1,000224.14
Lot NumberS04H044 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000224.30,000224.A1,000224.14
Lot NumberB22S002 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000224.30,000224.A1,000224.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel powder, -100 mesh, 99+% (metals basis excluding Co), Co 0.1% max, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.