missing translation for 'onlineSavingsMsg'
Learn More
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11319288

Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11319288
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11319288

Brand: Thermo Scientific Alfa Aesar 043214.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Form: Conductive dendritic powder

Conductive surface coatings, inks, adhesives, and sealants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray powder
Assay Percent Range 99.8% (Metals basis)
Quantity 25 g
Mesh Size -325 Mesh
Odor Odorless
Form Conductive Dendritic Powder
Chemical Name or Material Nickel

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU01I029 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog Number043214.22,043214.A3,043214.14,043214.36
Lot NumberY30H052 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043214.22,043214.A3,043214.14,043214.36
Lot NumberM14I016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043214.22,043214.A3,043214.14,043214.36
Lot NumberP01F005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043214.22,043214.A3,043214.14,043214.36
Lot NumberX05H007 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043214.22,043214.A3,043214.14,043214.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.