missing translation for 'onlineSavingsMsg'
Learn More
Nickel powder, APS 3-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11321318

Nickel powder, APS 3-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11321318
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11321318

Brand: Thermo Scientific Alfa Aesar 010256.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

UN Number UN3089
Quantity 100 g
Percent Purity 99.9%
Odor Odorless
Particle Size APS 3 to 7 Micron
Weight Apparent density: 1.8g/cm3 to 2.7g/cm3
Form Powder
Assay (metals basis)
Chemical Name or Material Nickel powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ06K031 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog Number010256.36,010256.A3,010256.22
Lot NumberT23K001 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number010256.36,010256.A3,010256.22
Lot NumberS27K020 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number010256.36,010256.A3,010256.22
Lot NumberS10K053 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog Number010256.36,010256.A3,010256.22
Lot NumberS23J036 Certificate TypeCertificate of Analysis Date03/07/2023 Catalog Number010256.36,010256.A3,010256,010256.22
compliance-icons compliance-icons
Product Identifier
  • Nickel powder
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Flammable solid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Nickel

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel powder, APS 3-7 micron, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.