missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel powder, spherical, -300 mesh, 99.8% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Silver to Gray |
Appearance | Silver gray powder of flakes |
Assay Percent Range | 99.8% (Metals basis) |
Quantity | 100 g |
Mesh Size | -300 Mesh |
Odor | Odorless |
Form | Spherical Powder |
Chemical Name or Material | Nickel |
Safety and Handling
Product Identifier
- Nickel powder
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Flammable solid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H228-Flammable solid.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Nickel
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'