Learn More
Nickel powder, spherical, -400 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
26.13 € - 622.10 €
Chemical Identifiers
CAS | 7440-02-0 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360298
|
Thermo Scientific Alfa Aesar
043704.14 |
25 g |
26.13 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11350298
|
Thermo Scientific Alfa Aesar
043704.22 |
100 g |
64.22 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11380298
|
Thermo Scientific Alfa Aesar
043704.36 |
500 g |
171.86 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370298
|
Thermo Scientific Alfa Aesar
043704.A3 |
2 kg |
622.10 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Specifications
7440-02-0 | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
−400 Mesh | |
Spherical Powder | |
Nickel powder |
Ni | |
UN3089 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
≥99.8% | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H372-H351-H228-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.
P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
H228-H317-H350-H372
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 231-111-4
missing translation for 'rtecsNumber' : QR5950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only