missing translation for 'onlineSavingsMsg'
Learn More

Nickel slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.98% (metals basis), Thermo Scientific Chemicals

47.03 € - 646.22 €

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]
View More Specs

Products 3
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11334398
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
042326.09
10 g
47.03 €
10g
Please to purchase this item. Need a web account? Register with us today!
11314398
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
042326.18
50 g
175.88 €
50g
Please to purchase this item. Need a web account? Register with us today!
11324398
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
042326.30
250 g
646.22 €
250g
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

7440-02-0
58.69
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]

Spezifikation

7440-02-0
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
99.98%
≈0.45g/slug
(metals basis)
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
58.69
CHEBI:28112
Odorless
Slug
Nickel slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

missing translation for 'einecsNumber' : 231-111-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.