missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Stainless Steel Tongs with PTFE Coated Tips

Get a grip on your samples with these Stainless Steel Tongs with PTFE Coated Tips.

Brand:  Cole-Parmer™ Essentials 06443-00

Product Code. 11725764

  • 54.80 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tips coated with PTFE Stainless steel tongs measures 9 in. long.
  • 3 in. of the tips are coated with PTFE.
Specifications

Specifications

Sample handling
1 Ea.
Grooved
Nickel, Stainless Steel
9 in.
PTFE
228.6 mm
Tongs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Essentials Stainless Steel Tongs with PTFE Coated Tips > Stainless Steel Tongs with PTFE Coated Tips; Length: 228.6 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.