Learn More
Nickel standard solution, for AAS, 1 mg/ml Ni in 2% HNO3, Thermo Scientific Chemicals
19.70 € - 60.70 €
Specifications
Chemical Name or Material | Nickel standard solution |
---|---|
Concentration | 990 to 1010μg/mL Ni in 2 to 5% HNO3 (20°C) |
Name Note | 1 mg/ml Ni in 2% HNO3 |
CAS | 7440-02-0, 13478-00-7, 7697-37-2, 7732-18-5 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10226780
|
Thermo Scientific Acros
196151000 |
100 mL | Plastic bottle |
19.70 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10583941
|
Thermo Scientific Acros
196155000 |
500 mL | Plastic bottle |
60.70 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Nickel standard solution | |
1 mg/ml Ni in 2% HNO3 | |
Plastic bottle | |
1.9, 96.9 | |
Ni | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
58.69 | |
1.0320g/mL | |
Soluble in water |
990 to 1010μg/mL Ni in 2 to 5% HNO3 (20°C) | |
7440-02-0, 13478-00-7, 7697-37-2, 7732-18-5 | |
Green | |
2.1, 97.1 | |
MFCD00011137 MFCD06798735 | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
AAS | |
1.032 | |
Crystals, Crystalline Powder and/or Chunks |
Safety and Handling
GHS H Statement
May cause cancer by inhalation. May damage the unborn child. Causes severe skin burns and eye damage. May produce an allergic reaction. Harmful to aquatic life with long lasting effects. May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep only in original container.
Danger
- Nickel standard solution, 1 mg/ml Ni in 2-5% HNO3
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.