missing translation for 'onlineSavingsMsg'
Learn More

Nickel sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11383807
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12035-72-2
Molecular Formula Ni3S2
Molecular Weight (g/mol) 240.20
MDL Number MFCD01711880
InChI Key YGHCWPXPAHSSNA-UHFFFAOYSA-N
Synonym nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432
PubChem CID 22833473
SMILES S=[Ni][Ni][Ni]=S

Specifications

Melting Point 797°C
Color Black
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water(0,00735g/L at 20°C).
IUPAC Name nickel;(sulfanylidene-$l^{4}-sulfanylidene)nickel
Formula Weight 240.19
Appearance Black powder
Chemical Name or Material Nickel sulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'