Learn More
Nickel sulfide, 99% (metals basis), Thermo Scientific Chemicals
207.00 € - 750.00 €
Chemical Identifiers
CAS | 12035-51-7 |
---|---|
Molecular Formula | NiS2 |
Molecular Weight (g/mol) | 122.81 |
MDL Number | MFCD00016267 |
InChI Key | HNDMNBUITRNJKK-UHFFFAOYSA-N |
Synonym | nickel iv sulphide |
PubChem CID | 131675689 |
IUPAC Name | disulfanylidenenickel |
SMILES | S=[Ni]=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15472317
|
Thermo Scientific Alfa Aesar
045498.06 |
5 g |
207.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482317
|
Thermo Scientific Alfa Aesar
045498.14 |
25 g | |||||||
Description
Nickel sulfide is used as a semiconductor material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12035-51-7 | |
122.81 | |
HNDMNBUITRNJKK-UHFFFAOYSA-N | |
131675689 | |
S=[Ni]=S |
NiS2 | |
MFCD00016267 | |
nickel iv sulphide | |
disulfanylidenenickel |
Specifications
12035-51-7 | |
NiS2 | |
UN3077 | |
Insoluble in water. | |
S=[Ni]=S | |
122.81 | |
99% | |
Nickel sulfide |
Powder | |
MFCD00016267 | |
nickel iv sulphide | |
HNDMNBUITRNJKK-UHFFFAOYSA-N | |
disulfanylidenenickel | |
131675689 | |
(metals basis) |
Safety and Handling
GHS H Statement
H350-H372-H341-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H341-H350-H372
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only