missing translation for 'onlineSavingsMsg'
Learn More

Nickel tin oxide dihydrate, Thermo Scientific Chemicals

97.59 € - 410.04 €

Chemical Identifiers

CAS 12035-38-0
Molecular Formula NiO3Sn
Molecular Weight (g/mol) 225.40
MDL Number MFCD00054011
InChI Key SHULAURIUQUJKN-UHFFFAOYSA-N
Synonym Nickel stannate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358248
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039452.22
100 g
97.59 €
100g
Please to purchase this item. Need a web account? Register with us today!
11368248
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039452.36
500 g
410.04 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12035-38-0
225.40
SHULAURIUQUJKN-UHFFFAOYSA-N
NiO3Sn
MFCD00054011
Nickel stannate

Specifications

12035-38-0
Powder
MFCD00054011
Insoluble in water.
225.40
Odorless
Green
NiO3Sn
Nickel stannate
SHULAURIUQUJKN-UHFFFAOYSA-N
261.43 (225.40 Anhydrous)
Nickel tin oxide dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350i-H372

missing translation for 'einecsNumber' : 234-824-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only