Learn More
Nicotinamide, 99%, Thermo Scientific Chemicals
34.47 € - 162.81 €
Chemical Identifiers
CAS | 98-92-0 |
---|---|
Molecular Formula | C6H6N2O |
Molecular Weight (g/mol) | 122.127 |
MDL Number | MFCD00006395 |
InChI Key | DFPAKSUCGFBDDF-UHFFFAOYSA-N |
Synonym | nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide |
PubChem CID | 936 |
ChEBI | CHEBI:17154 |
IUPAC Name | pyridine-3-carboxamide |
SMILES | C1=CC(=CN=C1)C(=O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498730
|
Thermo Scientific Alfa Aesar
A15970.22 |
100 g |
34.47 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498536
|
Thermo Scientific Alfa Aesar
A15970.30 |
250 g |
52.06 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488536
|
Thermo Scientific Alfa Aesar
A15970.0B |
1000 g |
162.81 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinamide is a building block (active component) of coenzymes nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phopshate (NADP). It may be used as a substrate of nicotinamide phosphoribosyltransferase (NAMPT, EC 2.4.2.12), the rate-limiting enzyme for NAD(+) biosynthesis and an important signaling molecule. It is an amide derivative of vitamin B3 and a PARP inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNicotinamide is a building block (active component) of coenzymes nicotinamide adenine dinucleotide (NAD) and nicotinamide adenine dinucleotide phopshate (NADP). It may be used as a substrate of nicotinamide phosphoribosyltransferase (NAMPT, EC 2.4.2.12), the rate-limiting enzyme for NAD(+) biosynthesis and an important signaling molecule. It is an amide derivative of vitamin B3 and a PARP inhibitor.
Solubility
Soluble in water, ethanol , glycerol, DMSO , methanol, butanol and chloroform. Slightly soluble in ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
98-92-0 | |
122.127 | |
DFPAKSUCGFBDDF-UHFFFAOYSA-N | |
936 | |
pyridine-3-carboxamide |
C6H6N2O | |
MFCD00006395 | |
nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide | |
CHEBI:17154 | |
C1=CC(=CN=C1)C(=O)N |
Specifications
98-92-0 | |
182°C (360°F) | |
Odorless | |
MFCD00006395 | |
14,6523 | |
DFPAKSUCGFBDDF-UHFFFAOYSA-N | |
pyridine-3-carboxamide | |
936 | |
122.13 | |
Nicotinamide |
128°C to 132°C | |
99% | |
C6H6N2O | |
383619 | |
nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide | |
C1=CC(=CN=C1)C(=O)N | |
122.127 | |
CHEBI:17154 | |
99% |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Warning
missing translation for 'rtecsNumber' : QS3675000
missing translation for 'tsca' : TSCA
- Nicotinamide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : nicotinamide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.