missing translation for 'onlineSavingsMsg'
Learn More

β-Nicotinamide Adenine Dinucleotide, Oxidized, Free Acid, 99%, MP Biomedicals™

Product Code. 11410222
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 53-84-9
Molecular Formula C21H27N7O14P2
Molecular Weight (g/mol) 663.43
MDL Number MFCD00150377
InChI Key BAWFJGJZGIEFAR-WIWLTUSXNA-N
Synonym nicotinamide adenine dinucleotide, nad+, diphosphopyridine nucleotide, nad-oxidized, nicotinamide-adenine dinucleotide, dpn-ox, beta-nicotinamide adenine dinucleotide, dpn+, nad, nad +
PubChem CID 15938971
ChEBI CHEBI:57540
IUPAC Name [[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 0.99
Quantity 5 g
Formula Weight 663.4 (anhydrous)
Physical Form Powder
Chemical Name or Material β-Nicotinamide Adenine Dinucleotide, Oxidized, Free Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'