Learn More
β-Nicotinamide mononucleotide, MedChemExpress
MedChemExpress β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.
79.65 € - 302.40 €
Specifications
Chemical Name or Material | β-Nicotinamide mononucleotide |
---|---|
CAS | 1094-61-7 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C11H15N2O8P |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478778
|
Medchem Express
HY-F0004 |
10 mg |
79.65 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16474697
|
Medchem Express
HY-F0004 |
50 mg |
191.70 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16468926
|
Medchem Express
HY-F0004 |
100 mg |
302.40 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
β-nicotinamide mononucleotide (β-NM) is a product of the nicotinamide phosphoribosyltransferase (NAMPT) reaction and a key NAD+ intermediate. The pharmacological activities of β-nicotinamide mononucleotide include its role in cellular biochemical functions, cardioprotection, diabetes, Alzheimer's disease, and complications associated with obesity.Specifications
β-Nicotinamide mononucleotide | |
White | |
C11H15N2O8P | |
H2O : 83.33 mg/mL (249.33 mM; Need ultrasonic) | |
334.22 | |
98.0% | |
Solid |
1094-61-7 | |
Research | |
β-NM NMN | |
NC(C1=C[N+]([C@@H]2O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]2O)=CC=C1)=O | |
334.22 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only