missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinoyl chloride hydrochloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.
Solubility
It decomposes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Nicotinoyl chloride hydrochloride was used in the preparation of 1-[5-O-tert-butyldimethylsilyl-2-deoxy-2-fluoro-3-O-(3-pyridylcarbonyl)-β-D-ribofuranosyl]uracil, bis(ethylenedithio)tetrathiafulvalene derivatives and 7,16-dinicotinoylated or 7,16-diisonicotinoylated compounds via reaction with tetraaza[14]annulene and its complexes.
Solubility
It decomposes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 149°C to 155°C |
Quantity | 10 g |
UN Number | UN3261 |
Beilstein | 3655772 |
Sensitivity | Moisture sensitive |
Solubility Information | It decomposes in water. |
Formula Weight | 178.02 |
Percent Purity | 98% |
Chemical Name or Material | Nicotinoyl chloride hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'