missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Red, 99%, pure

Product Code. 10464311 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 10464311

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7385-67-3 | C20H18N2O2 | 318.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.38
MDL Number MFCD00011639
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
Synonym Nile blue A oxazone
PubChem CID 65182
ChEBI CHEBI:52169
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31

Specifications

Melting Point 203.0°C to 205.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 27, II, 468
Merck Index 15, 6628
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in acetone, acetonitrile, chloroform, thf,, ethanol, dmso, cs2, benzene, nitrobenzene, pyridin, e, hexane, diethyl ether
Infrared Spectrum Authentic
IUPAC Name 9-(diethylamino)benzo[a]phenoxazin-5-one
Formula Weight 318.38
Percent Purity 99%
Grade Pure
Packaging Glass bottle
UV_Vis 98.5% min.
UV Spectrometry Complies
Color Maroon to Red
Quantity 100 mg
Physical Form Powder
Chemical Name or Material Nile Red, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.