missing translation for 'onlineSavingsMsg'
Learn More

Niobium boride, 99% (metals basis)

Product Code. p-7049043
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11367417

Brand: Thermo Scientific Alfa Aesar 012573.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12007-29-3 | BNb | 103.716 g/mol

Niobium boride is widely used in ceramic materials, electrode materials, thin film materials and refractory.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-29-3
Molecular Formula BNb
Molecular Weight (g/mol) 103.716
MDL Number MFCD00016236
InChI Key VDZMENNHPJNJPP-UHFFFAOYSA-N
Synonym niobium boride, niobium boride nbb, niobiumylidyneborane, niobium monoboride, columbium boride, columbium boride, cl
PubChem CID 82882
IUPAC Name boranylidyneniobium
SMILES B#[Nb]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 114.53
Percent Purity 99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Niobium boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.