Learn More
Niobium carbide, 99+% (metals basis), Thermo Scientific Chemicals
68.64 € - 228.14 €
Chemical Identifiers
CAS | 12069-94-2 |
---|---|
Molecular Formula | CNb |
Molecular Weight (g/mol) | 104.92 |
MDL Number | MFCD00016237 |
InChI Key | AADPBXLVMHIYCT-UHFFFAOYSA-N |
Synonym | niobium carbide nbc, methane-niobium 1:1, methane niobium, niobium carbide trace metals basis 25g |
PubChem CID | 159431 |
IUPAC Name | methane;niobium |
SMILES | [C-]#[Nb+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356518
|
Thermo Scientific Alfa Aesar
012147.14 |
25 g |
68.64 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346518
|
Thermo Scientific Alfa Aesar
012147.22 |
100 g |
228.14 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Niobium carbide is used in tool bits for cutting tools. Zirconium carbide and niobium carbide can be used as refractory coatings in nuclear reactors. It is used to produce arc-welding rods and in corrosion-resistant steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12069-94-2 | |
104.92 | |
AADPBXLVMHIYCT-UHFFFAOYSA-N | |
159431 | |
[C-]#[Nb+] |
CNb | |
MFCD00016237 | |
niobium carbide nbc, methane-niobium 1:1, methane niobium, niobium carbide trace metals basis 25g | |
methane;niobium |
Specifications
3500°C | |
White | |
99+% (metals basis) | |
MFCD00016237 | |
Insoluble in water. | |
[C-]#[Nb+] | |
104.92 | |
104.91 | |
Gray powder | |
Niobium carbide |
12069-94-2 | |
Powder | |
CNb | |
niobium carbide nbc, methane-niobium 1:1, methane niobium, niobium carbide trace metals basis 25g | |
AADPBXLVMHIYCT-UHFFFAOYSA-N | |
methane;niobium | |
159431 | |
Odorless | |
7.6 g/mL |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 235-117-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.