missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium Foil, 0.025 ± 0.015mm (0.001 ± 0.0006in) thick, 0.998, Thermo Scientific™
Click to view available options
Quantity:
20 x 100 cm
20 x 500 cm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Color | Silver to Gray |
Appearance | Silver gray foil |
Assay Percent Range | 99.80% |
Quantity | 20 x 100 cm |
Odor | Odorless |
Form | Foil |
Chemical Name or Material | Niobium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction