missing translation for 'onlineSavingsMsg'
Learn More

Niobium foil, 0.3mm (0.01in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 7440-03-1
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338598
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040668.FY
50 x 100 mm N/A
N/A
11328598
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040668.GK
100 x 200 mm N/A
N/A
11348598
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040668.HQ
200 x 500 mm N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-03-1
92.91
GUCVJGMIXFAOAE-UHFFFAOYSA-N
23936
niobium
Nb
MFCD00011126
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
CHEBI:33344
[Nb]

Specifications

7440-03-1
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
niobium
23936
99.8%
≈12.9g/50x100mm
(metals basis)
Nb
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
[Nb]
92.91
CHEBI:33344
Odorless
Foil
Niobium foil, 0.3mm (0.01 in.) thick, Annealed
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-113-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only