Learn More
Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis), Thermo Scientific Chemicals
Form: Approx.42.9g/50 x 100mm
124.62 € - 1510.52 €
Chemical Identifiers
CAS | 7440-03-1 |
---|---|
Molecular Formula | Nb |
Molecular Weight (g/mol) | 92.91 |
MDL Number | MFCD00011126 |
InChI Key | GUCVJGMIXFAOAE-UHFFFAOYSA-N |
Synonym | columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio |
PubChem CID | 23936 |
ChEBI | CHEBI:33344 |
IUPAC Name | niobium |
SMILES | [Nb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371318
|
Thermo Scientific Alfa Aesar
010257.FY |
50 x 100 mm |
124.62 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381318
|
Thermo Scientific Alfa Aesar
010257.GK |
100 x 200 mm |
357.78 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361318
|
Thermo Scientific Alfa Aesar
010257.HQ |
200 x 500 mm |
1510.52 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-03-1 | |
92.91 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
23936 | |
niobium |
Nb | |
MFCD00011126 | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
CHEBI:33344 | |
[Nb] |
Specifications
Silver to Gray | |
Silver gray foil | |
Nb | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
[Nb] | |
92.91 | |
CHEBI:33344 | |
Foil |
7440-03-1 | |
99.80% | |
MFCD00011126 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
niobium | |
23936 | |
Odorless | |
Niobium |
Safety and Handling
missing translation for 'einecsNumber' : 231-113-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.