missing translation for 'onlineSavingsMsg'
Learn More

Niobium(IV) oxide, 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11483810
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Niobium(IV) oxide is used as a reducing agent, which reduces carbon dioxide and sulfur dioxide to carbon and sulfur respectively. It finds application in dielectric material for electronic capacitors. It is also used for manufacturing metal niobium metal, in optical glass and in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12034-59-2
Molecular Formula NbO2
Molecular Weight (g/mol) 124.90
MDL Number MFCD00016242
InChI Key HFLAMWCKUFHSAZ-UHFFFAOYSA-N
Synonym niobium dioxide, niobium iv oxide, niobium oxide, columbium dioxide, niobium iv oxide trace metals basis, niobium iv oxide trace metals basis 10g
PubChem CID 82839
IUPAC Name dioxoniobium
SMILES O=[Nb]=O

Specifications

Physical Form Powder
Quantity 50 g
Assay Percent Range >99% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 124.91
Density 5.9 g/mL
Chemical Name or Material Niobium(IV) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'