missing translation for 'onlineSavingsMsg'
Learn More

Niobium phenoxide, Thermo Scientific™

Product Code. 11438650
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16576-63-9
Molecular Formula C30H25NbO5
Molecular Weight (g/mol) 558.431
MDL Number MFCD00014076
InChI Key ZCNDFHXTXLMYDM-UHFFFAOYSA-I
Synonym niobium 5+ pentakisphenoxide, pentaphenoxyniobium v, niobium 5+ ion pentaphenolate, columbium penta phenoxide, niobium 5+ pentaphenolate, phenol, niobium 5+ salt 5:1
PubChem CID 25021451
IUPAC Name niobium(5+);pentaphenoxide
SMILES C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].[Nb+5]

Specifications

Melting Point 208°C
Linear Formula Nb(C6H5O)5
Quantity 0.5 g
Sensitivity Moisture sensitive
Formula Weight 558.44
Physical Form Powder
Chemical Name or Material Niobium phenoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'