missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium powder, APS 1-5 micron, 99.8% (metals basis), Thermo Scientific Chemicals
52.06 € - 502.50 €
Chemical Identifiers
CAS | 7440-03-1 |
---|---|
Molecular Formula | Nb |
Molecular Weight (g/mol) | 92.91 |
MDL Number | MFCD00011126 |
InChI Key | GUCVJGMIXFAOAE-UHFFFAOYSA-N |
Synonym | columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio |
PubChem CID | 23936 |
ChEBI | CHEBI:33344 |
IUPAC Name | niobium |
SMILES | [Nb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11328458
|
Thermo Scientific Alfa Aesar
040478.09 |
10 g |
52.06 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318458
|
Thermo Scientific Alfa Aesar
040478.22 |
100 g |
262.31 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338458
|
Thermo Scientific Alfa Aesar
040478.30 |
250 g |
502.50 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in high temperature and super alloys of Zn and Ti
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-03-1 | |
92.91 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
23936 | |
niobium |
Nb | |
MFCD00011126 | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
CHEBI:33344 | |
[Nb] |
Specifications
Gray | |
gray powder | |
99.8% (Metals basis) | |
MFCD00011126 | |
GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
niobium | |
23936 | |
Odorless | |
Niobium |
7440-03-1 | |
8.57g/cm3 at 20°C | |
Nb | |
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio | |
[Nb] | |
92.91 | |
CHEBI:33344 | |
Powder |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: II
missing translation for 'einecsNumber' : 231-113-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only