missing translation for 'onlineSavingsMsg'
Learn More

Niobium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis excluding Ta), Thermo Scientific™

Product Code. 15484815
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Ea.
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 15484815

missing translation for 'mfr': Thermo Scientific Alfa Aesar 040885.KS

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3-1-7440
Quantity 1 Ea.
Odor Odorless
Molecular Formula Nb
MDL Number MFCD00011126
Synonym columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
Solubility Information Insoluble in water.
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES [Nb]
IUPAC Name niobium
Molecular Weight (g/mol) 92.91
PubChem CID 23936
ChEBI CHEBI:33344
Percent Purity 99.95%
Assay (metals basis excluding Ta)
Chemical Name or Material Niobium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.