Learn More
Niobium(V) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
134.00 € - 1175.00 €
Chemical Identifiers
CAS | 10026-12-7 |
---|---|
Molecular Formula | Cl5Nb |
Molecular Weight (g/mol) | 270.156 |
MDL Number | MFCD00011127 |
InChI Key | YHBDIEWMOMLKOO-UHFFFAOYSA-I |
Synonym | niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem |
PubChem CID | 24818 |
IUPAC Name | pentachloroniobium |
SMILES | Cl[Nb](Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11350338
|
Thermo Scientific Alfa Aesar
043729.06 |
5 g |
134.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370338
|
Thermo Scientific Alfa Aesar
043729.14 |
25 g |
407.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11360338
|
Thermo Scientific Alfa Aesar
043729.22 |
100 g |
1175.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Niobium(V) chloride is the precursor in the preparation of pure niobium, and ferroniobium. It is used as a Lewis acid catalyst in organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
10026-12-7 | |
270.156 | |
YHBDIEWMOMLKOO-UHFFFAOYSA-I | |
24818 | |
Cl[Nb](Cl)(Cl)(Cl)Cl |
Cl5Nb | |
MFCD00011127 | |
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem | |
pentachloroniobium |
Specifications
248.2°C | |
10026-12-7 | |
Cl5Nb | |
UN3260 | |
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem | |
YHBDIEWMOMLKOO-UHFFFAOYSA-I | |
pentachloroniobium | |
24818 | |
99.999% | |
Acidic | |
2.74 g/mL | |
Niobium (V) chloride |
208.3°C | |
Powder | |
MFCD00011127 | |
14,6557 | |
Soluble in alcohol,hydrochloric acid,carbon tetrachloride and chloroform. | |
Cl[Nb](Cl)(Cl)(Cl)Cl | |
270.156 | |
270.2 | |
Puratronic™ | |
Moisture sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 233-059-8
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only