missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) ethoxide, 99.95%, (trace metal basis), ACROS Organics™

153.00 € - 642.72 €

Chemical Identifiers

CAS 3236-82-6
Molecular Formula C10H25NbO5
Molecular Weight (g/mol) 318.21
MDL Number MFCD00015122
InChI Key ZTILUDNICMILKJ-UHFFFAOYSA-N
Synonym niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
PubChem CID 160675
SMILES CCO[Nb](OCC)(OCC)(OCC)OCC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10145900
View Documents
Thermo Scientific Acros
316880050
5 g Glass bottle
153.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
10627114
View Documents
Thermo Scientific Acros
316880500
50 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

3236-82-6
318.21
ZTILUDNICMILKJ-UHFFFAOYSA-N
160675
C10H25NbO5
MFCD00015122
niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
CCO[Nb](OCC)(OCC)(OCC)OCC

Specifications

140.0°C to 142.0°C (0.1mmHg)
3236-82-6
100.0
99.9%
Nb(OC2H5)5
36°C
Solubility in water: reacts
CCO[Nb](OCC)(OCC)(OCC)OCC
318.21
318.21
Trace Metal Basis
1.2680g/mL
6.0°C
99.9
Liquid
C10H25NbO5
MFCD00015122
niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+
ZTILUDNICMILKJ-UHFFFAOYSA-N
pentaethoxyniobium
160675
99.9%
1.268
Niobium(V) ethoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 221-795-2

Product Identifier
  • Niobium(V) ethoxide

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.