missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium(V) fluoride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Niobium(V) fluoride is used as a starting material to prepare other niobium compounds. It acts as an intermediate during the recovery of niobium metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | ∼230°C |
Melting Point | 80°C |
Color | White to Yellow |
Physical Form | Powder |
Quantity | 10 g |
UN Number | UN3260 |
Merck Index | 14,6558 |
Solubility Information | Hydrolyzes in water,alcohol,and caustic solutions; Sparingly soluble in CS2,chloroform; Forms complexes with Lewis basesSlightly soluble in carbon disulfide and chloroform. |
Formula Weight | 187.91 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'