missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) fluoride, 99%, Thermo Scientific Chemicals

Product Code. 11314959
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Niobium(V) fluoride is used as a starting material to prepare other niobium compounds. It acts as an intermediate during the recovery of niobium metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-68-8
Molecular Formula F5Nb
Molecular Weight (g/mol) 187.90
MDL Number MFCD00011129
InChI Key AOLPZAHRYHXPLR-UHFFFAOYSA-I
Synonym Niobium pentafluoride
IUPAC Name niobium(5+) pentafluoride
SMILES [F-].[F-].[F-].[F-].[F-].[Nb+5]

Specifications

Boiling Point ∼230°C
Melting Point 80°C
Color White to Yellow
Physical Form Powder
Quantity 50 g
UN Number UN3260
Merck Index 14,6558
Solubility Information Hydrolyzes in water,alcohol,and caustic solutions; Sparingly soluble in CS2,chloroform; Forms complexes with Lewis basesSlightly soluble in carbon disulfide and chloroform.
Formula Weight 187.91
Percent Purity 99%
Sensitivity Moisture sensitive
Density 3.92 g/mL
Chemical Name or Material Niobium (V) fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'