Learn More
Niobium(V) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
147.00 € - 542.00 €
Chemical Identifiers
CAS | 1313-96-8 |
---|---|
Molecular Formula | Nb2O5 |
Molecular Weight (g/mol) | 265.81 |
MDL Number | MFCD00011128 |
InChI Key | ZKATWMILCYLAPD-UHFFFAOYSA-N |
Synonym | niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde |
PubChem CID | 123105 |
IUPAC Name | dioxoniobiooxy(dioxo)niobium |
SMILES | O=[Nb](=O)O[Nb](=O)=O |
Description
Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in the food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators including linear and non-linear optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-96-8 | |
265.81 | |
ZKATWMILCYLAPD-UHFFFAOYSA-N | |
123105 | |
O=[Nb](=O)O[Nb](=O)=O |
Nb2O5 | |
MFCD00011128 | |
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde | |
dioxoniobiooxy(dioxo)niobium |
Specifications
1520°C | |
White | |
99.9% (metals basis) | |
MFCD00011128 | |
Insoluble in water. Soluble in hydrogen fluoride. | |
O=[Nb](=O)O[Nb](=O)=O | |
265.81 | |
265.81 | |
White powder or lump | |
Niobium(V) oxide |
1313-96-8 | |
Powder | |
Nb2O5 | |
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde | |
ZKATWMILCYLAPD-UHFFFAOYSA-N | |
dioxoniobiooxy(dioxo)niobium | |
123105 | |
Odorless | |
4.6 g/cm3 at 20°C |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'einecsNumber' : 215-213-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.