missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxide, 99.99%, (trace metal basis)

Product Code. 10153970 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10153970

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O

Specifications

Melting Point 1520.0°C
Color White
CAS Min % 99.95
CAS Max % 100.0
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99%
Linear Formula Nb2O5
Merck Index 15, 6645
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hf and hot h2so4, soluble in fused potassium hydrogen sulfate or, carbonates or hydroxides of the alkali metals
IUPAC Name [(dioxoniobio)oxy]dioxoniobium
Formula Weight 265.8
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 4.47
Density 4.4700g/mL
Chemical Name or Material Niobium(V) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.