missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

541.00 € - 1894.00 €

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396947
View Documents
Thermo Scientific Alfa Aesar
010822.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11316957
View Documents
Thermo Scientific Alfa Aesar
010822.A1
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products, and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators, including linear and non-linear optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1313-96-8
265.81
ZKATWMILCYLAPD-UHFFFAOYSA-N
123105
Nb2O5
MFCD00011128
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
O=[Nb](=O)O[Nb](=O)=O

Specifications

1520°C
White
99.9985% (metals basis)
MFCD00011128
Insoluble in water; soluble in hydrogen fluoride.
O=[Nb](=O)O[Nb](=O)=O
265.81
265.81
White powder or lump
Niobium(V) oxide
1313-96-8
Powder
Nb2O5
niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
ZKATWMILCYLAPD-UHFFFAOYSA-N
[(dioxoniobio)oxy]dioxoniobium
123105
Odorless
4.6 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'einecsNumber' : 215-213-6

missing translation for 'rtecsNumber' : QU0500000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.